Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ALDH1A1 Monoclonal Antibody (2B2G1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548364
Description
ALDH1A1 Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA
ALDH1A1 belongs to the aldehyde dehydrogenases family of proteins. Aldehyde dehydrogenase is the second enzyme of the major oxidative pathway of alcohol metabolism. Two major liver isoforms of this enzyme, cytosolic and mitochondrial, can be distinguished by their electrophoretic mobilities, kinetic properties, and subcellular localizations. Most Caucasians have two major isozymes, while approximately 50% of Orientals have only the cytosolic isozyme, missing the mitochondrial isozyme. A remarkably higher frequency of acute alcohol intoxication among Orientals than among Caucasians could be related to the absence of the mitochondrial isozyme.
Specifications
ALDH1A1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P00352 | |
ALDH1A1 | |
Purified recombinant fragment of human ALDH1A1 (AA: 1-110) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2B2G1 | |
Unconjugated | |
ALDH1A1 | |
acetaldehyde dehydrogenase 1; Ahd2; Ahd-2; alcohol dehydrogenase family 1, subfamily A1; alcohol dehydrogenase family 1, subfamily A2; ALDC; aldehyde dehydrogenase 1 family member A1; aldehyde dehydrogenase 1 family, member A1; Aldehyde dehydrogenase 1 soluble; aldehyde dehydrogenase 1, liver cytosolic (class 1); aldehyde dehydrogenase 1, soluble; aldehyde dehydrogenase 1, subfamily A1; aldehyde dehydrogenase 1A1; aldehyde dehydrogenase 2; aldehyde dehydrogenase family 1 member A1; aldehyde dehydrogenase family 1, member A1; aldehyde dehydrogenase family 1, subfamily A1; aldehyde dehydrogenase, cytosolic; aldehyde dehydrogenase, liver cytosolic; Aldh; ALDH class 1; Aldh1; ALDH11; ALDH1A1; Aldh1a2; Aldh2; ALDH-E1; ALHDII; E1; epididymis luminal protein 12; epididymis luminal protein 9; epididymis secretory sperm binding protein Li 53e; epididymis secretory sperm binding protein Li53e; HEL12; HEL-9; HEL-S-53e; MGC2318; PUMB1; RALDH 1; ralDH1; Retinal dehydrogenase 1; retinaldehyde dehydrogenase 1 | |
Mouse | |
Protein G | |
RUO | |
216 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction