Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALDH1A2 Antibody (SR2014), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | ALDH1A2 |
---|---|
Clone | SR2014 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
ALDH1A2 Monoclonal antibody specifically detects ALDH1A2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
ALDH1A2 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
aldehyde dehydrogenase 1 family, member A2, Aldehyde dehydrogenase family 1 member A2, EC 1.2.1, EC 1.2.1.36, RALDH 2, RALDH(II), RalDH2, RALDH2MGC26444, RALDH2-T, retinal dehydrogenase 2, Retinaldehyde-specific dehydrogenase type 2 | |
A synthesized peptide derived from human ALDH1A2 (Uniprot #: O94788) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2014 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology, Neuroscience | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
8854 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title