Learn More
Invitrogen™ ALDH1A2 Polyclonal Antibody

Description
Human ALDH1A2 shares 95.5% amino acid (aa) sequence identity with both mouse and rat ALDH1A2. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, human 293T whole cell, human HepG2 whole cell, human Hela whole cell, rat kidney tissue, rat testis tissue, mouse kidney tissue, mouse testis tissue. IHC: human colorectal adenocarcinoma tissue, rat lung tissue. Flow: 293T cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | ALDH1A2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and no preservative |
| Gene | ALDH1A2 |
| Gene Accession No. | O94788, Q62148, Q63639 |
| Gene Alias | AL1A2; alcohol dehydrogenase family 1, subfamily A2; alcohol dehydrogenase family 1, subfamily A7; aldehyde dehydrogenase 1 family member A2; aldehyde dehydrogenase 1 family, member A2; aldehyde dehydrogenase 1A2; aldehyde dehydrogenase family 1 member A2; aldehyde dehydrogenase family 1, subfamily A2; Aldh1a2; Aldh1a7; AV116159; fb50h01; neckless; nls; nof; no-fin; RALDH 2; RALDH(II); Raldh1; RALDH2; Raldh-2; RALDH2-T; retinal dehydrogenase 2; retinal dehydrogenase, type II; retinaldehyde dehydrogenase 2; retinaldehyde dehydrogenase type 2; retinaldehyde-specific dehydrogenase type 2 |
| Gene Symbols | ALDH1A2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.