Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH1A3 Monoclonal Antibody (OTI4B4), TrueMAB™, OriGene
SDP

Catalog No. 502541276
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1276 30 μL
50-167-3852 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1276 Supplier OriGene Supplier No. TA502842S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ALDH1A3 Monoclonal antibody specifically detects ALDH1A3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ALDH1A3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4B4
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ALDH1A3
Gene Accession No. P47895
Gene Alias acetaldehyde dehydrogenase 6; aldehyde dehydrogenase 1 family member A3; aldehyde dehydrogenase 1 family, member A3; aldehyde dehydrogenase 6; Aldehyde dehydrogenase family 1 member A3; aldehyde dehydrogenase family 1, subfamily A3; aldehyde dehydrogenase-6; ALDH1A3; ALDH1A6; ALDH6; ALDHA1A3; MCOP8; ralDH3; RALDH-3; Retinaldehyde dehydrogenase 3; V1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-100 and 413-512 of human ALDH1A3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.