Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ALDH1A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561201
Description
Immunogen sequence: ERGGAMATAN GAVENGQPDR KPPALPRPIR NLEVKFTKIF IN Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
ALDH1A3 are aldehyde dehydrogenase isozymes which are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. The enzyme encoded by the ALDH1A3 gene uses retinal as a substrate, either in a free or cellular retinol-binding protein form.
Specifications
ALDH1A3 | |
Polyclonal | |
Unconjugated | |
ALDH1A3 | |
acetaldehyde dehydrogenase 6; aldehyde dehydrogenase 1 family member A3; aldehyde dehydrogenase 1 family, member A3; aldehyde dehydrogenase 6; Aldehyde dehydrogenase family 1 member A3; aldehyde dehydrogenase family 1, subfamily A3; aldehyde dehydrogenase-6; ALDH1A3; ALDH1A6; ALDH6; ALDHA1A3; MCOP8; ralDH3; RALDH-3; Retinaldehyde dehydrogenase 3; V1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
220 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P47895 | |
ALDH1A3 | |
Recombinant protein corresponding to Human ALDH1A3. Recombinant protein control fragment (Product #RP-98180). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction