Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH1A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114838
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114838 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114838 Supplier Thermo Scientific Supplier No. PA5114838
Add to Cart
Add to Cart

ALDH1A3 Polyclonal Antibody for Western Blot, IHC (PFA)

TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH1A3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ALDH1A3
Gene Accession No. P47895, Q8K4D8, Q9JHW9
Gene Alias acetaldehyde dehydrogenase 6; aldehyde dehydrogenase 1 family member A3; aldehyde dehydrogenase 1 family, member A3; aldehyde dehydrogenase 6; Aldehyde dehydrogenase family 1 member A3; aldehyde dehydrogenase family 1, subfamily A3; aldehyde dehydrogenase-6; ALDH1A3; ALDH1A6; ALDH6; ALDHA1A3; MCOP8; ralDH3; RALDH-3; Retinaldehyde dehydrogenase 3; V1
Gene Symbols ALDH1A3
Host Species Rabbit
Immunogen A synthesized peptide derived from human ALDH1A3(Accession P47895), corresponding to amino acid residues E324-K374.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220, 266603, 56847
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.