Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH1B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109378
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109378 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109378 Supplier Invitrogen™ Supplier No. PA5109378
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLKRVTLELG GKSPSIVLAD ADMEHAVEQC HEALFFNMGQ CCCAGSRTFV EESIYNEFLE RTVEKAKQRK VGNPFELDTQ QGPQVDKEQF ERVLGYIQLG QKEGAKLLCG GERFGERGFF IKPTVFGGVQ DDMRIAKEEI FGPVQPLFKF KKIEEVVERA NNTRYGLAAA VFTRDLDKAM YFTQALQAGT VWVNTYNIVT CHTPFGGFKE SGNGRELGED GLKAYTEVKT VTIKVPQKNS.

Aldehyde dehydrogenases (ALDHs) mediate NADP+-dependent oxidation of aldehydes into acids during detoxification of alcohol-derived acetaldehyde, lipid peroxidation and metabolism of corticosteroids, biogenic amines and neurotransmitters. Alcohol drinking habits and cardiovascular disease risk factors may be associated with ALDH gene variants. ALDH1B1 (Aldehyde dehydrogenase family 1 member B1), also known as ALDH5 or ALDHX (Aldehyde dehydrogenase X, mitochondrial), is a 517 amino acid mitochondrial protein that is expressed in the liver, testis and to a lesser extent in brain. ALDH1B1 belongs to the aldehyde dehydrogenase family and may play a major role in ethanol detoxification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH1B1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ALDH1B1
Gene Accession No. P30837, Q66HF8, Q9CZS1
Gene Alias 2700007F14Rik; acetaldehyde dehydrogenase 5; aldehyde dehydrogenase 1 family member B1; aldehyde dehydrogenase 1 family, member B1; aldehyde dehydrogenase 1B1; aldehyde dehydrogenase 5; Aldehyde dehydrogenase family 1 member B1; aldehyde dehydrogenase X, mitochondrial; ALDH class 2; ALDH1B1; ALDH5; ALDHX; MGC2230
Gene Symbols ALDH1B1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 278-517 of human ALDH1B1 (NP_000683.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219, 298079, 72535
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.