Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALDH1L1 Antibody (2E7), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250033AF532
Description
ALDH1L1 Monoclonal antibody specifically detects ALDH1L1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
ALDH1L1 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
2E7 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
10-formyltetrahydrofolate dehydrogenase, aldehyde dehydrogenase 1 family, member L1, aldehyde dehydrogenase family 1 member L1, Cytosolic 10-formyltetrahydrofolate dehydrogenase, DKFZp781N0997, EC 1.5.1.6,10-FTHFDH, FDH, formyltetrahydrofolate dehydrogenase, FTHFD10-fTHF | |
Amino acids 402-902 of human ALDH1L1. [UniProt# O75891] | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Endocrinology, Neuroscience, Signal Transduction | |
10840 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction