Learn More
Invitrogen™ ALDH1L1 Monoclonal Antibody (7G8), eBioscience™
Mouse Monoclonal Antibody
$427.00 - $1562.00
Specifications
Antigen | ALDH1L1 |
---|---|
Clone | 7G8 |
Concentration | 0.5 mg/mL |
Content And Storage | 4°C |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
14-959-537
![]() |
Invitrogen™
14959537 |
2 mg |
Each for $1,562.00
|
|
|||||
50-112-2793
![]() |
Invitrogen™
14959582 |
100 μg |
Each for $427.00
|
|
|||||
Description
Description: This 7G8 monoclonal antibody reacts with human 10-formyltetrahydrofolate dehydrogenase (ALDH1L1) a protein involved in folate metabolism. The cytosolic enzyme, with homology to the aldehyde dehydrogenase (ALDH) family, is involved in the conversion of 10-formyltetrahydrofolate, NADP and water to tetrahydrofolate, NADPH and carbon dioxide. Folate metabolism is essential for cell growth and proliferation and decreased levels of folate have been implicated in neural tube defects in addition to increased cancer risk. This isoform (1L1) is expressed in glial cells has been identified as a highly specific marker for astrocytes labeling both the cell body and processes. Applications Reported: This 7G8 antibody has been reported for use in flow cytometric analysis, western blotting, immunohistochemical staining, and immunocytochemistry. Applications Tested: This 7G8 antibody has been tested by immunohistochemistry on human FFPE tissue using both high and low pH antigen retrieval. Both antigen retrieval methods resulted in staining although low pH antigen retrieval is optimal. The antibody can be used at less than or equal to 1 μg/mL. It is recommended that anibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
ALDH1L1 catalyzes the conversion of 10-formyltetrahydrofolate, NADP, and water to tetrahydrofolate, NADPH, and carbon dioxide. The encoded protein belongs to the aldehyde dehydrogenase family and is responsible for formate oxidation in vivo. Deficiencies in this gene can result in an accumulation of formate and subsequent methanol poisoning.Specifications
ALDH1L1 | |
0.5 mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
O75891 | |
10840 | |
Primary | |
Affinity chromatography |
7G8 | |
4°C | |
Monoclonal | |
Liquid | |
IgG2b κ | |
Human | |
ALDH1L1 | |
10-formyltetrahydrofolate dehydrogenase; 10fTHF; 10-fTHF; 10FTHFDH; 10-FTHFDH; 1810048F20Rik; aldehyde dehydrogenase 1 family member L1; aldehyde dehydrogenase 1 family, member L1; aldehyde dehydrogenase family 1 member L1; Aldh1l1; Cytosolic 10-formyltetrahydrofolate dehydrogenase; DKFZp781N0997; FBP-CI; FDH; formyltetrahydrofolate dehydrogenase; Fthfd; LOC100855730; MER57B-ALDH1L1; Neut2 | |
ALDH1L1 | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.