Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH1L1 Monoclonal Antibody (OTI6A10), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA501871

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-2978


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

ALDH1L1 catalyzes the conversion of 10-formyltetrahydrofolate, NADP, and water to tetrahydrofolate, NADPH, and carbon dioxide. The encoded protein belongs to the aldehyde dehydrogenase family and is responsible for formate oxidation in vivo. Deficiencies in this gene can result in an accumulation of formate and subsequent methanol poisoning.
Specifications

Specifications

ALDH1L1
Monoclonal
0.69 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O75891
ALDH1L1, LOC100855730
Full length human recombit protein of human ALDH1L1 produced in HEK293T cell.
100 μL
Primary
Canine, Human
Antibody
IgG1
Flow Cytometry, Western Blot
OTI6A10
Unconjugated
ALDH1L1
10-FTHFDH, 10-fTHF, FDH, FTHFD
Mouse
Affinity Chromatography
RUO
100855730, 10840
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.