Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH1L1 Monoclonal Antibody (OTI7B7), TrueMAB™, OriGene
SDP

Catalog No. 502541964
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1964 30 μL
50-167-2965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1964 Supplier OriGene Supplier No. TA501855S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ALDH1L1 Monoclonal antibody specifically detects ALDH1L1 in Canine, Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen ALDH1L1
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI7B7
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ALDH1L1
Gene Accession No. O75891
Gene Alias 10-formyltetrahydrofolate dehydrogenase; 10fTHF; 10-fTHF; 10FTHFDH; 10-FTHFDH; 1810048F20Rik; aldehyde dehydrogenase 1 family member L1; aldehyde dehydrogenase 1 family, member L1; aldehyde dehydrogenase family 1 member L1; Aldh1l1; Cytosolic 10-formyltetrahydrofolate dehydrogenase; DKFZp781N0997; FBP-CI; FDH; formyltetrahydrofolate dehydrogenase; Fthfd; LOC100855730; MER57B-ALDH1L1; Neut2
Host Species Mouse
Immunogen Full length human recombinant protein of human ALDH1L1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100855730, 10840
Target Species Canine, Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.