Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH1L1 Recombinant Rabbit Monoclonal Antibody (JU53-54)
GREENER_CHOICE

Catalog No. PIMA541249
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541249 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA541249 Supplier Invitrogen™ Supplier No. MA541249
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

ALDH1L1 Recombinant Monoclonal Antibody for IHC (F), IHC (P), Western Blot

ALDH1L1 catalyzes the conversion of 10-formyltetrahydrofolate, NADP, and water to tetrahydrofolate, NADPH, and carbon dioxide. The encoded protein belongs to the aldehyde dehydrogenase family and is responsible for formate oxidation in vivo. Deficiencies in this gene can result in an accumulation of formate and subsequent methanol poisoning.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH1L1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JU53-54
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene ALDH1L1
Gene Accession No. O75891, P28037, Q8R0Y6
Gene Alias 10-formyltetrahydrofolate dehydrogenase; 10fTHF; 10-fTHF; 10FTHFDH; 10-FTHFDH; 1810048F20Rik; aldehyde dehydrogenase 1 family member L1; aldehyde dehydrogenase 1 family, member L1; aldehyde dehydrogenase family 1 member L1; Aldh1l1; Cytosolic 10-formyltetrahydrofolate dehydrogenase; DKFZp781N0997; FBP-CI; FDH; formyltetrahydrofolate dehydrogenase; Fthfd; LOC100855730; MER57B-ALDH1L1; Neut2
Gene Symbols ALDH1L1
Host Species Rabbit
Immunogen Recombinant protein within Human ALDH1L1 aa 155-375.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107747, 10840, 64392
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.