Learn More
Invitrogen™ ALDH1L2 Polyclonal Antibody

Description
Immunogen sequence: FGNDGKALTV RNLQFEDGKM IPASQYFSTG ETSVVELTAE EVKVAETIKV IWAGILSNVP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.
Specifications
Specifications
| Antigen | ALDH1L2 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ALDH1L2 |
| Gene Accession No. | Q3SY69 |
| Gene Alias | 10-formyltetrahydrofolate dehydrogenase ALDH1L2; aldehyde dehydrogenase 1 family member L2; aldehyde dehydrogenase 1 family, member L2; aldehyde dehydrogenase 1L2; Aldehyde dehydrogenase family 1 member L2; aldehyde dehydrogenase family 1 member L2, mitochondrial; ALDH1L2; D330038I09Rik; mitochondrial 10-formyltetrahydrofolate dehydrogenase; Mitochondrial 10-FTHFDH; mtFDH; probable 10-formyltetrahydrofolate dehydrogenase ALDH1L2 |
| Gene Symbols | ALDH1L2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.