Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH1L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584407
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584407 Supplier Invitrogen™ Supplier No. PA584407
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KCGGLQLQNE DVYMATKFEG FIQKVVRKLR GEDQEVELVV DYISKEVNEI MVKMPYQCFI NGQFTDADDG KT Highest antigen sequence indentity to the following orthologs: Rat - 85%, Mouse - 79%.

This gene encodes a member of both the aldehyde dehydrogenase superfamily and the formyl transferase superfamily. This member is the mitochondrial form of 10-formyltetrahydrofolate dehydrogenase (FDH), which converts 10-formyltetrahydrofolate to tetrahydrofolate and CO2 in an NADP(+)-dependent reaction, and plays an essential role in the distribution of one-carbon groups between the cytosolic and mitochondrial compartments of the cell. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH1L2
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALDH1L2
Gene Accession No. Q3SY69
Gene Alias 10-formyltetrahydrofolate dehydrogenase ALDH1L2; aldehyde dehydrogenase 1 family member L2; aldehyde dehydrogenase 1 family, member L2; aldehyde dehydrogenase 1L2; Aldehyde dehydrogenase family 1 member L2; aldehyde dehydrogenase family 1 member L2, mitochondrial; ALDH1L2; D330038I09Rik; mitochondrial 10-formyltetrahydrofolate dehydrogenase; Mitochondrial 10-FTHFDH; mtFDH; probable 10-formyltetrahydrofolate dehydrogenase ALDH1L2
Gene Symbols ALDH1L2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALDH1L2. Recombinant protein control fragment (Product #RP-108438).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160428
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.