Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH1L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120273
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120273 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120273 Supplier Invitrogen™ Supplier No. PA5120273
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse pancreas, Rat pancreas, Rat liver. The target is usually found in the following locations: Mitochondrion.

This gene encodes a member of both the aldehyde dehydrogenase superfamily and the formyl transferase superfamily. This member is the mitochondrial form of 10-formyltetrahydrofolate dehydrogenase (FDH), which converts 10-formyltetrahydrofolate to tetrahydrofolate and CO2 in an NADP(+)-dependent reaction, and plays an essential role in the distribution of one-carbon groups between the cytosolic and mitochondrial compartments of the cell. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH1L2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ALDH1L2
Gene Accession No. Q3SY69, Q8K009
Gene Alias 10-formyltetrahydrofolate dehydrogenase ALDH1L2; aldehyde dehydrogenase 1 family member L2; aldehyde dehydrogenase 1 family, member L2; aldehyde dehydrogenase 1L2; Aldehyde dehydrogenase family 1 member L2; aldehyde dehydrogenase family 1 member L2, mitochondrial; ALDH1L2; D330038I09Rik; mitochondrial 10-formyltetrahydrofolate dehydrogenase; Mitochondrial 10-FTHFDH; mtFDH; probable 10-formyltetrahydrofolate dehydrogenase ALDH1L2
Gene Symbols ALDH1L2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160428, 216188, 299699
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.