Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALDH2 Antibody (2C10), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259488IR
This item is not returnable.
View return policy
Description
ALDH2 Monoclonal antibody specifically detects ALDH2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
ALDH2 | |
Monoclonal | |
DyLight 755 | |
acetaldehyde dehydrogenase 2, aldehyde dehydrogenase 2 family (mitochondrial), aldehyde dehydrogenase, mitochondrial, ALDH class 2, ALDH-E2, ALDHI, ALDM, EC 1.2.1, EC 1.2.1.3, liver mitochondrial ALDH, MGC1806, nucleus-encoded mitochondrial aldehyde dehydrogenase 2 | |
Recombinant human ALDH2 (18-517aa) purified from E. coli | |
0.1 mL | |
Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
217 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
2C10 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction