Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529717
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529717 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529717 Supplier Invitrogen™ Supplier No. PA529717
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, HeLa, HepG2, mouse brain. Predicted reactivity: Mouse (97%), Rat (96%), Xenopus laevis (83%), Pig (92%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ALDH2 belongs to the aldehyde dehydrogenase family of proteins. Aldehyde dehydrogenase is the second enzyme of the major oxidative pathway of alcohol metabolism. Two major liver isoforms of this enzyme, cytosolic and mitochondrial, can be distinguished by their electrophoretic mobilities, kinetic properties, and subcellular localizations. Most Caucasians have two major isozymes, while approximately 50% of Asians have only the cytosolic isozyme, missing the mitochondrial isozyme. A remarkably higher frequency of acute alcohol intoxication among Asians than among Caucasians could be related to the absence of the mitochondrial isozyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Aldh2
Gene Accession No. P05091, P11884, P47738
Gene Alias acetaldehyde dehydrogenase 2; Ahd1; Ahd-1; Ahd5; Ahd-5; AHD-M1; aldehyde dehydrogenase 2 family (mitochondrial); aldehyde dehydrogenase 2, mitochondrial; aldehyde dehydrogenase, mitochondrial; ALDH class 2; ALDH1; ALDH2; ALDH-E2; ALDHI; ALDM; EC 1.2.1.3; liver mitochondrial ALDH; MGC1806; mitochondrial aldehyde dehydrogenase; nucleus-encoded mitochondrial aldehyde dehydrogenase 2
Gene Symbols Aldh2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ALDH2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11669, 217, 29539
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.