Learn More
Invitrogen™ ALDH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588450
Description
Immunogen sequence: AGSRTFVQED IYDEFVERSV ARAKSRVVGN PFDSKTEQGP QVDETQFKKI LGYINTGKQE GAKLLCGGGI AADRGYFIQP TVFGDVQDGM TIAKEEIFGP VMQILKFKTI EEVVGRANNS TYGLAAAVFT KDLDKANYLS QALQAGTVWV NCYDVFGAQS Positive Samples: Mouse lung, Mouse liver; Cellular Location: Mitochondrion matrix.
ALDH2 belongs to the aldehyde dehydrogenase family of proteins. Aldehyde dehydrogenase is the second enzyme of the major oxidative pathway of alcohol metabolism. Two major liver isoforms of this enzyme, cytosolic and mitochondrial, can be distinguished by their electrophoretic mobilities, kinetic properties, and subcellular localizations. Most Caucasians have two major isozymes, while approximately 50% of Asians have only the cytosolic isozyme, missing the mitochondrial isozyme. A remarkably higher frequency of acute alcohol intoxication among Asians than among Caucasians could be related to the absence of the mitochondrial isozyme.
Specifications
ALDH2 | |
Polyclonal | |
Unconjugated | |
Aldh2 | |
acetaldehyde dehydrogenase 2; Ahd1; Ahd-1; Ahd5; Ahd-5; AHD-M1; aldehyde dehydrogenase 2 family (mitochondrial); aldehyde dehydrogenase 2, mitochondrial; aldehyde dehydrogenase, mitochondrial; ALDH class 2; ALDH1; ALDH2; ALDH-E2; ALDHI; ALDM; EC 1.2.1.3; liver mitochondrial ALDH; MGC1806; mitochondrial aldehyde dehydrogenase; nucleus-encoded mitochondrial aldehyde dehydrogenase 2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
11669, 217, 29539 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.06 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P05091, P11884, P47738 | |
Aldh2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 321-480 of human ALDH2 (NP_0006812). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.