Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH2 Polyclonal Antibody, Huabio

Rabbit Polyclonal Antibody

Supplier:  HUABIO RESEARCH INC R14073

Encompass
Encompass_Preferred

Catalog No. 50-254-4402


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

ALDH2 Polyclonal antibody specifically detects ALDH2 in Human samples. It is validated for Immunocytochemistry, Western Blot
Specifications

Specifications

ALDH2
Polyclonal
Unconjugated
Aldh2
acetaldehyde dehydrogenase 2; Ahd1; Ahd-1; Ahd5; Ahd-5; AHD-M1; aldehyde dehydrogenase 2 family (mitochondrial); aldehyde dehydrogenase 2, mitochondrial; aldehyde dehydrogenase, mitochondrial; ALDH class 2; ALDH1; ALDH2; ALDH-E2; ALDHI; ALDM; EC 1.2.1.3; liver mitochondrial ALDH; MGC1806; mitochondrial aldehyde dehydrogenase; nucleus-encoded mitochondrial aldehyde dehydrogenase 2
Synthetic peptide within human ALDH2 aa 18-80.
100 μL
Primary
Human
Antibody
IgG
Immunocytochemistry, Western Blot
1 mg/mL
PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
P05091
Rabbit
Antigen affinity chromatography
RUO
217
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.