Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH3A1 Monoclonal Antibody (OTI1C6)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525098

Catalog No. PIMA525098


Only null left
Add to Cart

Description

Description

ALDH3A1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

Aldh3A1 is a member of the aldehyde dehydrogenase superfamily, a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of aliphatic and aromatic aldehydes. Aldh3A1 is highly expressed in stomach and even more strongly in cornea, representing between 5 to 50% of the water soluble protein fraction in mammalian corneas. It is thought that Aldh3A1 acts to protect the cornea from UV-induced oxidative stress by not only detoxification of reactive aldehydes by also through the direct absorption of UV energy. However, corneas from Aldh3A1-null mice are indistinguishable from those from wild-type mice; mice lacking both Aldh3A1 and Aldh1A1 showed increased cataract formation following UVB exposure, suggesting that Aldh1A1 may be able to compensate for the loss of Aldh3A1.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ALDH3A1
Monoclonal
0.96 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P30838
ALDH3A1
Full length human recombinant protein of ALDH3A1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2a
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI1C6
Unconjugated
ALDH3A1
Ahd4; Ahd-4; AHDC; Ahd-c; alcohol dehydrogenase family 3, subfamily A1; Aldd; Aldehyde dehydrogenase (Ahd-c); Aldehyde dehydrogenase 3; aldehyde dehydrogenase 3 family member A1; aldehyde dehydrogenase 3 family, member A1; aldehyde dehydrogenase 3 family, memberA1; aldehyde dehydrogenase 3, stomach cytosolic (class 3); aldehyde dehydrogenase 3A1; aldehyde dehydrogenase 4; aldehyde dehydrogenase class 3; aldehyde dehydrogenase family 3 member A1; aldehyde dehydrogenase family 3 subfamily A1; aldehyde dehydrogenase family 3, member A1; aldehyde dehydrogenase family 3, subfamily A1; aldehyde dehydrogenase isozyme 3; aldehyde dehydrogenase type III; aldehyde dehydrogenase, dimeric NADP-preferring; Aldh; Aldh3; Aldh3a1; Aldh4; ALDHIII; BCP54; bovine corneal protein 54; corneal 15.8 kDa protein; corneal epithelium; corneal protein 54, transparentin; dehydrogenase 3, dimeric NAPD-prefering; dioxin-inducible aldehyde dehydrogenase 3; HTC-ALDH; MGC10406; stomach aldehyde dehydrogenase; transparentin; Tumor-associated aldehyde dehydrogenase; tumor-associated aldehyde dehydrogenase tumor ALDH
Mouse
Affinity Chromatography
RUO
218
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.