Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH3A1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983823
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3823 Supplier Bioss Supplier No. BS15496R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aldh3A1 is a member of the aldehyde dehydrogenase superfamily, a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of aliphatic and aromatic aldehydes. Aldh3A1 is highly expressed in stomach and even more strongly in cornea, representing between 5 to 50% of the water soluble protein fraction in mammalian corneas. It is thought that Aldh3A1 acts to protect the cornea from UV-induced oxidative stress by not only detoxification of reactive aldehydes by also through the direct absorption of UV energy. However, corneas from Aldh3A1-null mice are indistinguishable from those from wild-type mice; mice lacking both Aldh3A1 and Aldh1A1 showed increased cataract formation following UVB exposure, suggesting that Aldh1A1 may be able to compensate for the loss of Aldh3A1.

Specifications

Antigen ALDH3A1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ALDH3A1
Gene Accession No. P30838
Gene Alias Ahd4; Ahd-4; AHDC; Ahd-c; alcohol dehydrogenase family 3, subfamily A1; Aldd; Aldehyde dehydrogenase (Ahd-c); Aldehyde dehydrogenase 3; aldehyde dehydrogenase 3 family member A1; aldehyde dehydrogenase 3 family, member A1; aldehyde dehydrogenase 3 family, memberA1; aldehyde dehydrogenase 3, stomach cytosolic (class 3); aldehyde dehydrogenase 3A1; Aldehyde dehydrogenase 4; aldehyde dehydrogenase class 3; Aldehyde dehydrogenase family 3 member A1; aldehyde dehydrogenase family 3 subfamily A1; aldehyde dehydrogenase family 3, member A1; aldehyde dehydrogenase family 3, subfamily A1; aldehyde dehydrogenase isozyme 3; aldehyde dehydrogenase type III; Aldehyde dehydrogenase, dimeric NADP-preferring; Aldh; ALDH3; Aldh3a1; Aldh4; ALDHIII; BCP54; BOS_18909; corneal 15.8 kDa protein; corneal protein 54, transparentin; dehydrogenase 3, dimeric NAPD-prefering; Dioxin-inducible aldehyde dehydrogenase 3; HTC-ALDH; MGC10406; stomach aldehyde dehydrogenase; transparentin; Tumor-associated aldehyde dehydrogenase; tumor-associated aldehyde dehydrogenase tumor ALDH
Gene Symbols ALDH3A1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ALDH3A1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.