Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH3A1 Recombinant Rabbit Monoclonal Antibody (316)
GREENER_CHOICE

Catalog No. PIMA529025
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529025 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529025 Supplier Invitrogen™ Supplier No. MA529025
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human ALDH3A1.

Aldh3A1 is a member of the aldehyde dehydrogenase superfamily, a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of aliphatic and aromatic aldehydes. Aldh3A1 is highly expressed in stomach and even more strongly in cornea, representing between 5 to 50% of the water soluble protein fraction in mammalian corneas. It is thought that Aldh3A1 acts to protect the cornea from UV-induced oxidative stress by not only detoxification of reactive aldehydes by also through the direct absorption of UV energy. However, corneas from Aldh3A1-null mice are indistinguishable from those from wild-type mice; mice lacking both Aldh3A1 and Aldh1A1 showed increased cataract formation following UVB exposure, suggesting that Aldh1A1 may be able to compensate for the loss of Aldh3A1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH3A1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 316
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ALDH3A1
Gene Accession No. P30838
Gene Alias Ahd4; Ahd-4; AHDC; Ahd-c; alcohol dehydrogenase family 3, subfamily A1; Aldd; Aldehyde dehydrogenase (Ahd-c); Aldehyde dehydrogenase 3; aldehyde dehydrogenase 3 family member A1; aldehyde dehydrogenase 3 family, member A1; aldehyde dehydrogenase 3 family, memberA1; aldehyde dehydrogenase 3, stomach cytosolic (class 3); aldehyde dehydrogenase 3A1; aldehyde dehydrogenase 4; aldehyde dehydrogenase class 3; aldehyde dehydrogenase family 3 member A1; aldehyde dehydrogenase family 3 subfamily A1; aldehyde dehydrogenase family 3, member A1; aldehyde dehydrogenase family 3, subfamily A1; aldehyde dehydrogenase isozyme 3; aldehyde dehydrogenase type III; aldehyde dehydrogenase, dimeric NADP-preferring; Aldh; Aldh3; Aldh3a1; Aldh4; ALDHIII; BCP54; bovine corneal protein 54; corneal 15.8 kDa protein; corneal epithelium; corneal protein 54, transparentin; dehydrogenase 3, dimeric NAPD-prefering; dioxin-inducible aldehyde dehydrogenase 3; HTC-ALDH; MGC10406; stomach aldehyde dehydrogenase; transparentin; Tumor-associated aldehyde dehydrogenase; tumor-associated aldehyde dehydrogenase tumor ALDH
Gene Symbols ALDH3A1
Host Species Rabbit
Immunogen Recombinant Human ALDH3A1 Protein (Met1-His453).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.