Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALDH3A2 Monoclonal Antibody (OTI1H10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503143S


Description
ALDH3A2 Monoclonal antibody specifically detects ALDH3A2 in Human samples. It is validated for Western BlotSpecifications
ALDH3A2 | |
Monoclonal | |
0.52 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
A1L0T0 | |
Mouse | |
Affinity chromatography | |
RUO | |
10994 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Western Blot | |
OTI1H10 | |
Unconjugated | |
ILVBL | |
Ahd3; Ahd-3; Ahd-3r; Ahd3-r; AI194803; alcohol dehydrogenase family 3, subfamily A2; aldehyde dehydrogenase 10; Aldehyde dehydrogenase 3; aldehyde dehydrogenase 3 family member A2; aldehyde dehydrogenase 3 family, member A2; aldehyde dehydrogenase 3A2; aldehyde dehydrogenase 4; Aldehyde dehydrogenase family 3 member A2; aldehyde dehydrogenase family 3 member A2; fatty aldehyde dehydrogenase; aldehyde dehydrogenase family 3, subfamily A2; ALDH10; Aldh3; Aldh3a2; Aldh4; Aldh4-r; DKFZp686E23276; FALDH; fatty aldehyde dehydrogenase; Fatty aldehyde dehydrogenase-like protein; FLJ20851; microsomal aldehyde dehydrogenase; msALDH; QccE-15682; SLS | |
Full length human recombinant protein of human ILVBL produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction