Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH3A2 Mouse anti-Human, Clone: 151CT1.3.1, Abnova™

Catalog No. 89933549 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-549 Supplier Abnova Corporation Supplier No. MAB12297
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human ALDH3A2.

Aldehyde dehydrogenase isozymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. This gene product catalyzes the oxidation of long-chain aliphatic aldehydes to fatty acid. Mutations in the gene cause Sjogren-Larsson syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen ALDH3A2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 151CT1.3.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human ALDH3A2.
Dilution Immunohistochemistry (1:50-1:100) Western Blot (1:200-1:10000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.09% sodium azide)
Gene Alias ALDH10/DKFZp686E23276/FALDH/FLJ20851/SLS
Gene Symbols ALDH3A2
Host Species Mouse
Immunogen Recombinant protein corresponding to human ALDH3A2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.