Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH3B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101411
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101411 Supplier Thermo Scientific Supplier No. PA5101411
Add to Cart
Add to Cart

ALDH3B1 Polyclonal Antibody

ALDH3B1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH3B1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ALDH3B1
Gene Accession No. P43353
Gene Alias 1700001N19Rik; aldehyde dehydrogenase 3 family member B1; aldehyde dehydrogenase 3 family, member B1; aldehyde dehydrogenase 3B1; aldehyde dehydrogenase 7; Aldehyde dehydrogenase family 3 member B1; Aldh3b1; ALDH4; ALDH7; FLJ26433; FLJ34710
Gene Symbols ALDH3B1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ALDH3B1(Accession P43353), corresponding to amino acid residues L30-A80.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.