Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH3B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558425
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558425 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558425 Supplier Invitrogen™ Supplier No. PA558425
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AAQLQGLGRF LQENKQLLHD ALAQDLHKSA FESEVSEVAI SQGEVTLALR NLRAWMKDER VPKNLATQLD SAFIRKEPF Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 84%.

The aldehyde dehydrogenases are a family of isozymes that may play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. This particular gene spans about 20 kb of genomic DNA and is composed of 9 coding exons. The gene is highly expressed in kidney and lung. The functional significance of this gene as well as the cellular localization of its product are presently unknown. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH3B1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALDH3B1
Gene Accession No. P43353
Gene Alias 1700001N19Rik; aldehyde dehydrogenase 3 family member B1; aldehyde dehydrogenase 3 family, member B1; aldehyde dehydrogenase 3B1; aldehyde dehydrogenase 7; Aldehyde dehydrogenase family 3 member B1; Aldh3b1; ALDH4; ALDH7; FLJ26433; FLJ34710
Gene Symbols ALDH3B1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALDH3B1. Recombinant protein control fragment (Product #RP-97264).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.