Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH3B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537122
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537122 Supplier Invitrogen™ Supplier No. PA537122
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 43 kDa. Purity is >95% by SDS-PAGE.

ALDH3B2 (aldehyde dehydrogenase 3 family, member B2), also known as ALDH8, is a 385 amino acid protein that belongs to the ALDH family and is involved in the pathway of alcohol metabolism. Expressed in salivary gland tissue, ALDH3B2 functions to catalyze the NADP+-dependent conversion of an aldehyde into an acid. The gene encoding ALDH3B2 maps to human chromosome 11, which houses over 1, 400 genes and comprises nearly 4% of the human genome. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are associated with defects in genes that maps to chromosome 11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH3B2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Aldh3b2
Gene Accession No. E9Q3E1, P48448
Gene Alias acetaldehyde dehydrogenase 8; AI848594; aldehyde dehydrogenase 3 family member B2; aldehyde dehydrogenase 3 family, member B2; aldehyde dehydrogenase 8; Aldehyde dehydrogenase family 3 member B2; Aldh3b2; ALDH8; C130048D07Rik
Gene Symbols Aldh3b2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues in human ALDH3B2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222, 621603, 688800
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.