Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ALDH3B2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560934
Description
Immunogen sequence: SGLEKLKEIH YPPYTDWNQQ LLRWGMGSQS CTLL Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 68%.
ALDH3B2 (aldehyde dehydrogenase 3 family, member B2), also known as ALDH8, is a 385 amino acid protein that belongs to the ALDH family and is involved in the pathway of alcohol metabolism. Expressed in salivary gland tissue, ALDH3B2 functions to catalyze the NADP+-dependent conversion of an aldehyde into an acid. The gene encoding ALDH3B2 maps to human chromosome 11, which houses over 1, 400 genes and comprises nearly 4% of the human genome. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are associated with defects in genes that maps to chromosome 11.
Specifications
ALDH3B2 | |
Polyclonal | |
Unconjugated | |
Aldh3b2 | |
acetaldehyde dehydrogenase 8; AI848594; aldehyde dehydrogenase 3 family member B2; aldehyde dehydrogenase 3 family, member B2; aldehyde dehydrogenase 8; Aldehyde dehydrogenase family 3 member B2; Aldh3b2; ALDH8; C130048D07Rik | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
222 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P48448 | |
Aldh3b2 | |
Recombinant protein corresponding to Human ALDH3B2. Recombinant protein control fragment (Product #RP-100382). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction