Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALDH4A1 Antibody (001), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290151MFV500
Description
ALDH4A1 Monoclonal antibody specifically detects ALDH4A1 in Human samples. It is validated for Flow CytometrySpecifications
ALDH4A1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
8659 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
001 | |
Flow Cytometry | |
aldehyde dehydrogenase 4 family, member A1, Aldehyde dehydrogenase family 4 member A1, ALDH4DKFZp779M035, delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial, EC 1.5.1.12, mitochondrial delta-1-pyrroline 5-carboxylate dehydrogenase, P5C dehydrogenase, P5CD, P5CDh | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ALDH4A1 (AAH07581.1; Lys25-Gln563). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction