Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALDH4A1 Antibody [Alexa Fluor« 532], Novus Biologicals Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP338007AF532
Description
ALDH4A1 Polyclonal antibody specifically detects ALDH4A1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
ALDH4A1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
8659 | |
Store at 4°C in the dark. | |
Purified |
ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
Alexa Fluor 532 | |
aldehyde dehydrogenase 4 family, member A1, Aldehyde dehydrogenase family 4 member A1, ALDH4DKFZp779M035, delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial, EC 1.5.1.12, mitochondrial delta-1-pyrroline 5-carboxylate dehydrogenase, P5C dehydrogenase, P5CD, P5CDh | |
Recombinant fusion protein containing a sequence corresponding to amino acids 354-563 of human ALDH4A1 (NP_003739.2).,, Sequence:, LYVPHSLWPQIKGRLLEEHSRIKVGDPAEDFGTFFSAVIDAKSFARIKKWLEHARSSPSLTILAGGKCDDSVGYFVEPCIVESKDPQEPIMKEEIFGPVLSVYVYPDDKYKETLQLVDSTTSYGLTGAVFSQDKDVVQEATKVLRNAAGNFYINDKSTGSIVGQQPFGGARASGTNDKPGGPHYILRWTSPQVIKETHKPLGDWSYAYMQ | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction