Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH4A1 Monoclonal Antibody (7E3)
GREENER_CHOICE

Catalog No. PIMA534908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534908 Supplier Invitrogen™ Supplier No. MA534908
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: SK-Br-3, HepG2, human liver tissue, human colon cancer tissue, human prostate tissue, human kidney tissue.

ALDH4A1 belongs to the aldehyde dehydrogenase family of proteins. This enzyme is a mitochondrial matrix NAD-dependent dehydrogenase which catalyzes the second step of the proline degradation pathway, converting pyrroline-5-carboxylate to glutamate. Deficiency of this enzyme is associated with type II hyperprolinemia, an autosomal recessive disorder characterized by accumulation of delta-1-pyrroline-5-carboxylate (P5C) and proline.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH4A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7E3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene Aldh4a1
Gene Accession No. P30038
Gene Alias A930035F14Rik; Ahd1; Ahd-1; aldehyde dehydrogenase 4 family member A1; aldehyde dehydrogenase 4 family, member A1; aldehyde dehydrogenase 4A1; aldehyde dehydrogenase family 4 member A1; ALDH4; aldh4a1; Aldh5a1; delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial; E330022C09; L-glutamate gamma-semialdehyde dehydrogenase; mitochondrial delta-1-pyrroline 5-carboxylate dehydrogenase; P5C dehydrogenase; P5cd; P5CDh; P5cdhl; P5cdhs; RP11-128M10.1; similar to aldehyde dehydrogenase 4 family, member A1; Ssdh1; zgc:63592
Gene Symbols Aldh4a1
Host Species Mouse
Immunogen Synthetic peptide within Human ALDH4A1 aa 514-563.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8659
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.