Learn More
ALDH4A1 Mouse anti-Canine, Human, Rat, Clone: OTI1H10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF812719

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This protein belongs to the aldehyde dehydrogenase family of proteins. This enzyme is a mitochondrial matrix NAD-dependent dehydrogenase which catalyzes the second step of the proline degradation pathway, converting pyrroline-5-carboxylate to glutamate. Deficiency of this enzyme is associated with type II hyperprolinemia, an autosomal recessive disorder characterized by accumulation of delta-1-pyrroline-5-carboxylate (P5C) and proline. Two transcript variants encoding the same protein have been identified for this gene.Specifications
ALDH4A1 | |
Monoclonal | |
Unconjugated | |
ALDH4A1 | |
ALDH4, P5CD, P5CDh | |
Mouse | |
Affinity Chromatography | |
RUO | |
612452, 641316, 8659 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1H10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P0C2X9, P30038 | |
ALDH4A1 | |
Human recombit protein fragment corresponding to amino acids 1-503 of human ALDH4A1 produced in E.coli. | |
100 μg | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.