Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH4A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102639
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102639 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102639 Supplier Invitrogen™ Supplier No. PA5102639
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ALDH4A1.

ALDH4A1 belongs to the aldehyde dehydrogenase family of proteins. This enzyme is a mitochondrial matrix NAD-dependent dehydrogenase which catalyzes the second step of the proline degradation pathway, converting pyrroline-5-carboxylate to glutamate. Deficiency of this enzyme is associated with type II hyperprolinemia, an autosomal recessive disorder characterized by accumulation of delta-1-pyrroline-5-carboxylate (P5C) and proline.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH4A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Aldh4a1
Gene Accession No. P0C2X9, P30038, Q8CHT0
Gene Alias A930035F14Rik; Ahd1; Ahd-1; aldehyde dehydrogenase 4 family member A1; aldehyde dehydrogenase 4 family, member A1; aldehyde dehydrogenase 4A1; aldehyde dehydrogenase family 4 member A1; ALDH4; aldh4a1; Aldh5a1; delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial; E330022C09; L-glutamate gamma-semialdehyde dehydrogenase; mitochondrial delta-1-pyrroline 5-carboxylate dehydrogenase; P5C dehydrogenase; P5cd; P5CDh; P5cdhl; P5cdhs; RP11-128M10.1; similar to aldehyde dehydrogenase 4 family, member A1; Ssdh1; zgc:63592
Gene Symbols Aldh4a1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ALDH4A1(Accession P30038), corresponding to amino acid residues N499-E549.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212647, 641316, 8659
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.