Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH4A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529119
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529119 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529119 Supplier Invitrogen™ Supplier No. PA529119
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2. Predicted reactivity: Mouse (90%), Rat (90%), Pig (91%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ALDH4A1 belongs to the aldehyde dehydrogenase family of proteins. This enzyme is a mitochondrial matrix NAD-dependent dehydrogenase which catalyzes the second step of the proline degradation pathway, converting pyrroline-5-carboxylate to glutamate. Deficiency of this enzyme is associated with type II hyperprolinemia, an autosomal recessive disorder characterized by accumulation of delta-1-pyrroline-5-carboxylate (P5C) and proline.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH4A1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Aldh4a1
Gene Accession No. P30038
Gene Alias A930035F14Rik; Ahd1; Ahd-1; aldehyde dehydrogenase 4 family member A1; aldehyde dehydrogenase 4 family, member A1; aldehyde dehydrogenase 4A1; aldehyde dehydrogenase family 4 member A1; ALDH4; aldh4a1; Aldh5a1; delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial; E330022C09; L-glutamate gamma-semialdehyde dehydrogenase; mitochondrial delta-1-pyrroline 5-carboxylate dehydrogenase; P5C dehydrogenase; P5cd; P5CDh; P5cdhl; P5cdhs; RP11-128M10.1; similar to aldehyde dehydrogenase 4 family, member A1; Ssdh1; zgc:63592
Gene Symbols Aldh4a1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ALDH4A1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8659
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.