Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALDH5A1 Antibody (17F5), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259409UV
This item is not returnable.
View return policy
Description
ALDH5A1 Monoclonal antibody specifically detects ALDH5A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
ALDH5A1 | |
Monoclonal | |
DyLight 350 | |
aldehyde dehydrogenase 5 family, member A1, EC 1.2.1, EC 1.2.1.24, NAD(+)-dependent succinic semialdehyde dehydrogenase, SSADHmitochondrial | |
Recombinant human ALDH5A1 (48-535aa) purified from E. coli | |
0.1 mL | |
metabolism, Neuroscience | |
7915 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
17F5 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction