Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALDH5A1 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298042PE
Description
ALDH5A1 Polyclonal antibody specifically detects ALDH5A1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
ALDH5A1 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
aldehyde dehydrogenase 5 family, member A1, EC 1.2.1, EC 1.2.1.24, NAD(+)-dependent succinic semialdehyde dehydrogenase, SSADHmitochondrial | |
Produced in rabbits immunized with E. coli-derived Human ALDH5A1 fragment. | |
0.1 mL | |
metabolism, Neuroscience | |
7915 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction