Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ALDH5A1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519394
Description
This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
Aldh5A1 is a member of the aldehyde dehydrogenase superfamily, a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of aliphatic and aromatic aldehydes. Aldehyde dehydrogenase enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. Aldh5A1 is a mitochondrial NAD(+)-dependent succinic semialdehyde dehydrogenase. A deficiency of this enzyme, known as 4-hydroxybutyricaciduria, results in a disorder of the neurotransmitter 4-aminobutyric acid (GABA). Symptoms usually include static encephalopathy, associated with developmental delays, hypotonia, ataxia, speech defects, and seizures. At least two isoforms of Aldh5A1 are known to exist.
Specifications
| ALDH5A1 | |
| Polyclonal | |
| Unconjugated | |
| ALDH5A1 | |
| 6330403E24Rik; Ahd1; Ahd-1; aldehyde dehydrogenase 1, mitochondrial; aldehyde dehydrogenase 5 family member A1; aldehyde dehydrogenase 5 family, member A1; aldehyde dehydrogenase 5 family, member A1 (succinate-semialdehyde dehydrogenase); aldehyde dehydrogenase family 5 member A1; aldehyde dehydrogenase family 5, subfamily A1; ALDH5A1; aldhehyde dehydrogenase family 5, subfamily A1; D630032B01Rik; mitochondrial succinate semialdehyde dehydrogenase; NAD(+)-dependent succinic semialdehyde dehydrogenase; OTTMUSG00000000613; SSADH; SSDH; Ssdh1; Succinate-semialdehyde dehydrogenase, mitochondrial; succinic semialdehyde dehydrogenase | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 214579, 291133, 7915 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P51649, P51650, Q8BWF0 | |
| ALDH5A1 | |
| Synthetic peptide sequence (TGKILLHHAANS) corresponding to the internal amino acids of ALDH5A1 (aa 288-299). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction