Learn More
Invitrogen™ ALDH5A1 Polyclonal Antibody

Description
A suggested positive control is mouse liver tissue lysate. PA5-20609 can be used with blocking peptide PEP-0729.
Specifications
Specifications
| Antigen | ALDH5A1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ALDH5A1 |
| Gene Accession No. | P51649, P51650, Q8BWF0 |
| Gene Alias | 6330403E24Rik; Ahd1; Ahd-1; aldehyde dehydrogenase 1, mitochondrial; aldehyde dehydrogenase 5 family member A1; aldehyde dehydrogenase 5 family, member A1; aldehyde dehydrogenase 5 family, member A1 (succinate-semialdehyde dehydrogenase); aldehyde dehydrogenase family 5 member A1; aldehyde dehydrogenase family 5, subfamily A1; ALDH5A1; aldhehyde dehydrogenase family 5, subfamily A1; D630032B01Rik; mitochondrial succinate semialdehyde dehydrogenase; NAD(+)-dependent succinic semialdehyde dehydrogenase; OTTMUSG00000000613; SSADH; SSDH; Ssdh1; Succinate-semialdehyde dehydrogenase, mitochondrial; succinic semialdehyde dehydrogenase |
| Gene Symbols | ALDH5A1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.