Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH5A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597453
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597453 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597453 Supplier Invitrogen™ Supplier No. PA597453
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ALDH5A1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Aldh5A1 is a member of the aldehyde dehydrogenase superfamily, a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of aliphatic and aromatic aldehydes. Aldehyde dehydrogenase enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. Aldh5A1 is a mitochondrial NAD(+)-dependent succinic semialdehyde dehydrogenase. A deficiency of this enzyme, known as 4-hydroxybutyricaciduria, results in a disorder of the neurotransmitter 4-aminobutyric acid (GABA). Symptoms usually include static encephalopathy, associated with developmental delays, hypotonia, ataxia, speech defects, and seizures. At least two isoforms of Aldh5A1 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH5A1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ALDH5A1
Gene Accession No. P51649, P51650, Q8BWF0
Gene Alias 6330403E24Rik; Ahd1; Ahd-1; aldehyde dehydrogenase 1, mitochondrial; aldehyde dehydrogenase 5 family member A1; aldehyde dehydrogenase 5 family, member A1; aldehyde dehydrogenase 5 family, member A1 (succinate-semialdehyde dehydrogenase); aldehyde dehydrogenase family 5 member A1; aldehyde dehydrogenase family 5, subfamily A1; ALDH5A1; aldhehyde dehydrogenase family 5, subfamily A1; D630032B01Rik; mitochondrial succinate semialdehyde dehydrogenase; NAD(+)-dependent succinic semialdehyde dehydrogenase; OTTMUSG00000000613; SSADH; SSDH; Ssdh1; Succinate-semialdehyde dehydrogenase, mitochondrial; succinic semialdehyde dehydrogenase
Gene Symbols ALDH5A1
Host Species Rabbit
Immunogen Recombinant Human Succinate-semialdehyde dehydrogenase, mitochondrial protein (48-535AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214579, 291133, 7915
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.