Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH6A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530090
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530090 Supplier Invitrogen™ Supplier No. PA530090
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat. Predicted reactivity: Mouse (93%), Rat (92%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This protein belongs to the aldehyde dehydrogenases family of proteins. This enzyme plays a role in the valine and pyrimidine catabolic pathways. The product of this gene, a mitochondrial methylmalonate semialdehyde dehydrogenase, catalyzes the irreversible oxidative decarboxylation of malonate and methylmalonate semialdehydes to acetyl- and propionyl-CoA. Methylmalonate semialdehyde dehydrogenase deficiency is characterized by elevated beta-alanine, 3-hydroxypropionic acid, and both isomers of 3-amino and 3-hydroxyisobutyric acids in urine organic acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH6A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene ALDH6A1
Gene Accession No. Q02252
Gene Alias 1110038I05Rik; AI314632; aldehyde dehydrogenase 6 family member A1; aldehyde dehydrogenase 6 family, member A1; aldehyde dehydrogenase family 6 member A1; aldehyde dehydrogenase family 6, subfamily A1; Aldh6a1; malonate-semialdehyde dehydrogenase; methylmalonate semialdehyde dehydrogenase; methylmalonate-semialdehyde dehydrogenase [acylating], mitochondrial; MGC40271; mitochondrial acylating methylmalonate-semialdehyde dehydrogenase; MMSADHA; Mmsdh; testicular tissue protein Li 122
Gene Symbols ALDH6A1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ALDH6A1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4329
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.