Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALDH7A1 Antibody (101), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289926JF549
Description
ALDH7A1 Monoclonal specifically detects ALDH7A1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
ALDH7A1 | |
Monoclonal | |
Janelia Fluor 549 | |
aldehyde dehydrogenase 7 family, member A1, Aldehyde dehydrogenase family 7 member A1,26g turgor protein homolog, Alpha-AASA dehydrogenase, ATQ1Antiquitin-1, Betaine aldehyde dehydrogenase, Delta1-piperideine-6-carboxylate dehydrogenase, EC 1.2.1, EC 1.2.1.3, EC 1.2.1.31, EC 1.2.1.8, EPDalpha-aminoadipic semialdehyde dehydrogenase, FLJ11738, FLJ92814, P6c dehydrogenase, PDEantiquitin-1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ALDH7A1 (P49419-2; Ser2-Gln511). | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
501 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, Immunofluorescence | |
101 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction