Learn More
Invitrogen™ ALDH7A1 Monoclonal Antibody (OTI10A12)

Description
ALDH7A1 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | ALDH7A1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI10A12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ALDH7A1 |
| Gene Accession No. | P49419 |
| Gene Alias | 26g turgor protein homolog; Ald7a1; aldehyde dehydrogenase 7 family member A1; aldehyde dehydrogenase 7 family, member A1; aldehyde dehydrogenase family 7 member A1; aldehyde dehydrogenase family 7, member A1; Aldh7a1; Alpha-AASA dehydrogenase; alpha-aminoadipic semialdehyde dehydrogenase; antiquitin; Antiquitin 1; Antiquitin1; Antiquitin-1; ATQ1; Betaine aldehyde dehydrogenase; D18Wsu181e; delta1-piperideine-6-carboxylate dehydrogenase; delta1-piperideine-6-carboxylate dehydrogenease; EPD; P6c dehydrogenase; PDE |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.