Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH7A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529974
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529974 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529974 Supplier Invitrogen™ Supplier No. PA529974
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, H1299, HeLa, HepG2, mouse liver, rat liver. Predicted reactivity: Human (99%), Mouse (91%), Rat (92%), Zebrafish (82%), Xenopus laevis (81%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a member of subfamily 7 in the aldehyde dehydrogenase gene family. These enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. This particular member has homology to a previously described protein from the green garden pea, the 26g pea turgor protein. It is also involved in lysine catabolism that is known to occur in the mitochondrial matrix. Recent reports show that this protein is found both in the cytosol and the mitochondria, and the two forms likely arise from the use of alternative translation initiation sites. An additional variant encoding a different isoform has also been found for this gene. Mutations in this gene are associated with pyridoxine-dependent epilepsy. Several related pseudogenes have also been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH7A1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ALDH7A1
Gene Accession No. P49419, Q64057, Q9DBF1
Gene Alias 26g turgor protein homolog; Ald7a1; aldehyde dehydrogenase 7 family member A1; aldehyde dehydrogenase 7 family, member A1; aldehyde dehydrogenase family 7 member A1; aldehyde dehydrogenase family 7, member A1; Aldh7a1; Alpha-AASA dehydrogenase; alpha-aminoadipic semialdehyde dehydrogenase; antiquitin; Antiquitin 1; Antiquitin1; Antiquitin-1; ATQ1; Betaine aldehyde dehydrogenase; D18Wsu181e; delta1-piperideine-6-carboxylate dehydrogenase; delta1-piperideine-6-carboxylate dehydrogenease; EPD; P6c dehydrogenase; PDE
Gene Symbols ALDH7A1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ALDH7A1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110695, 291450, 501
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.