Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH8A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555298
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555298 Supplier Invitrogen™ Supplier No. PA555298
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPCSSYIDSY DPSTGEVYCR VPNSGKDEIE AAVKAAREAF PSWSSRSPQE RSRVLNQVAD LLEQS Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.

This protein belongs to the aldehyde dehydrogenases family of proteins. It plays a role in a pathway of 9-cis-retinoic acid biosynthesis in vivo. This enzyme converts 9-cis-retinal into the retinoid X receptor ligand 9-cis-retinoic acid, and has approximately 40-fold higher activity with 9-cis-retinal than with all-trans-retinal. Therefore, it is the first known aldehyde dehydrogenase to show a preference for 9-cis-retinal relative to all-trans-retinal. Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH8A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Aldh8a1
Gene Accession No. Q9H2A2
Gene Alias 2-aminomuconic semialdehyde dehydrogenase; Aldehyde dehydrogenase 12; aldehyde dehydrogenase 8 family member A1; aldehyde dehydrogenase 8 family, member A1; aldehyde dehydrogenase family 8 member A1; aldehyde dehydrogenase family protein; ALDH12; Aldh8a1; DJ352A20.2; Raldh4; Retinal dehydrogenase 4; retinal dehydrogenase type 4
Gene Symbols Aldh8a1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALDH8A1. Recombinant protein control fragment (Product #RP-94718).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64577
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.