Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALDH8A1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299392AF647
Description
ALDH8A1 Polyclonal specifically detects ALDH8A1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ALDH8A1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Aldehyde dehydrogenase 12, aldehyde dehydrogenase 8 family, member A1, aldehyde dehydrogenase family protein, ALDH12aldehyde dehydrogenase family 8 member A1, DJ352A20.2, DKFZp779D2315, EC 1.2.1, EC 1.2.1.-, EC 1.2.1.3, MGC138650 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human ALDH8A1. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
64577 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction