Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH9A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552756 Supplier Invitrogen™ Supplier No. PA552756
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DCDMNNAVKG ALMANFLTQG QVCCNGTRVF VQKEILDKFT EEVVKQTQRI KIGDPLLEDT RMGPLINRPH LERVLGFVKV AKEQGAKVLC GGDIYVPEDP KLKDGYYMRP CVLTNCRDDM TCVKEEIFGP VMSILSFD Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.

ALDH9A1 belongs to the aldehyde dehydrogenase family of proteins. The protein has a high activity for oxidation of gamma-aminobutyraldehyde and other amino aldehydes. The enzyme catalyzes the dehydrogenation of gamma-aminobutyraldehyde to gamma-aminobutyric acid (GABA). This isozyme is a tetramer of identical 54-kD subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH9A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALDH9A1
Gene Accession No. P49189
Gene Alias 4-trimethylaminobutyraldehyde dehydrogenase; 4-trimethylaminobutyraldehyde dehydrogenase, N-terminally processed; AA139417; Abaldh; aldehyde dehydrogenase (NAD+); aldehyde dehydrogenase 9 family member A1; aldehyde dehydrogenase 9 family, member A1; aldehyde dehydrogenase 9, subfamily A1; aldehyde dehydrogenase 9A1; aldehyde dehydrogenase E3 isozyme; Aldehyde dehydrogenase family 9 member A1; aldehyde dehydrogenase family 9, subfamily A1; ALDH4; ALDH7; ALDH9; Aldh9a1; E3; ESTM40; gamma-aminobutyraldehyde dehydrogenase; gamma-trimethylaminobutyraldehyde dehydrogenase; R-aminobutyraldehyde dehydrogenase; TMABADH; TMABA-DH
Gene Symbols ALDH9A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALDH9A1. Recombinant protein control fragment (Product #RP-89160).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.