Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH9A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596840
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596840 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596840 Supplier Invitrogen™ Supplier No. PA596840
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, 22Rv1, NCI-H460, Mouse liver, Mouse kidney, Rat liver, Rat kidney; Cellular Location: Cytoplasm Immunogen sequence: MFLRAGLAAL SPLLRSLRPS PVAAMSTGTF VVSQPLNYRG GARVEPADAS GTEKAFEPAT GRVIATFTCS GEKEVNLAVQ NAKAAFKIWS QKSGMERCRI LLEAARIIRE REDEIATMEC INNGKSIFEA RLDIDISWQC LEYYAGLAAS MAGEHIQLPG GSFGYTRREP LGVCVGIGAW.

ALDH9A1 belongs to the aldehyde dehydrogenase family of proteins. The protein has a high activity for oxidation of gamma-aminobutyraldehyde and other amino aldehydes. The enzyme catalyzes the dehydrogenation of gamma-aminobutyraldehyde to gamma-aminobutyric acid (GABA). This isozyme is a tetramer of identical 54-kD subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH9A1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ALDH9A1
Gene Accession No. P49189, Q9JLJ2, Q9JLJ3
Gene Alias 4-trimethylaminobutyraldehyde dehydrogenase; 4-trimethylaminobutyraldehyde dehydrogenase, N-terminally processed; AA139417; Abaldh; aldehyde dehydrogenase (NAD+); aldehyde dehydrogenase 9 family member A1; aldehyde dehydrogenase 9 family, member A1; aldehyde dehydrogenase 9, subfamily A1; aldehyde dehydrogenase 9A1; aldehyde dehydrogenase E3 isozyme; Aldehyde dehydrogenase family 9 member A1; aldehyde dehydrogenase family 9, subfamily A1; ALDH4; ALDH7; ALDH9; Aldh9a1; E3; ESTM40; gamma-aminobutyraldehyde dehydrogenase; gamma-trimethylaminobutyraldehyde dehydrogenase; R-aminobutyraldehyde dehydrogenase; TMABADH; TMABA-DH
Gene Symbols ALDH9A1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human ALDH9A1 (NP_0006873).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223, 56752, 64040
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.