Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aldo-keto Reductase 1C3/AKR1C3 Antibody (SR1612), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Aldo-keto Reductase 1C3/AKR1C3 |
---|---|
Clone | SR1612 |
Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:30 |
Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
Aldo-keto Reductase 1C3/AKR1C3 Monoclonal antibody specifically detects Aldo-keto Reductase 1C3/AKR1C3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
Aldo-keto Reductase 1C3/AKR1C3 | |
Western Blot 1:500-1:1000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:30 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
17-beta-hydroxysteroid dehydrogenase type 5, 3-alpha-HSD type 2, 3-alpha-hydroxysteroid dehydrogenase type 2, aldo-keto reductase family 1 member C3, aldo-keto reductase family 1, member C3 (3-alpha hydroxysteroid dehydrogenase, type II), Chlordecone reductase homolog HAKRb, DD-3, DD317-beta-HSD 5, DDH1, DDX, Dihydrodiol dehydrogenase 3, Dihydrodiol dehydrogenase type I, dihydrodiol dehydrogenase X, EC 1.1.1, EC 1.1.1.112,3-alpha-HSD type II, brain, EC 1.1.1.188, EC 1.1.1.213, EC 1.1.1.63, EC 1.1.1.64, EC 1.3.1.20, HA1753, HAKRBtype IIb 3-alpha hydroxysteroid dehydrogenase, HAKRe, HSD17B5, hydroxysteroid (17-beta) dehydrogenase 5, Indanol dehydrogenase, KIAA0119hluPGFS, PGFS, Prostaglandin F synthase, Testosterone 17-beta-dehydrogenase 5, Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase | |
A synthesized peptide derived from human Aldo-keto Reductase 1C3/AKR1C3 (Uniprot #: P42330) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1612 | |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
8644 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title