Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALDOA Mouse anti-Human, Clone:2H2B1, Abnova™
Mouse monoclonal antibody raised against partial recombinant human ALDOA.
Supplier: Abnova Corporation MAB17947
Description
This gene product, Aldolase A (fructose-bisphosphate aldolase) is a glycolytic enzyme that catalyzes the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate and dihydroxyacetone phosphate. Three aldolase isozymes (A, B, and C), encoded by three different genes, are differentially expressed during development. Aldolase A is found in the developing embryo and is produced in even greater amounts in adult muscle. Aldolase A expression is repressed in adult liver, kidney and intestine and similar to aldolase C levels in brain and other nervous tissue. Aldolase A deficiency has been associated with myopathy and hemolytic anemia. Alternative splicing of this gene results in multiple transcript variants which encode the same protein. [provided by RefSeq]Specifications
ALDOA | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Flow Cytometry (1:200-1:400) Immunofluorescence (1:100-1:500) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
ALDOA | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunofluorescence, Western Blot | |
2H2B1 | |
aldolase A, fructose-bisphosphate | |
In PBS (0.05% sodium azide) | |
ALDA, MGC10942, MGC17716, MGC17767 | |
Recombinant protein corresponding to amino acids 9-145 of human ALDOA. | |
226 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction