Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALDOB Monoclonal Antibody (OTI 5D1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
$231.00 - $516.29
Specifications
Antigen | ALDOB |
---|---|
Clone | OTI 5D1 |
Concentration | 0.84 mg/mL |
Applications | Flow Cytometry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-254-1141
![]() ![]() ![]() |
OriGene
TA503062S |
30 μL |
Each for $231.00
|
|
|||||
50-167-4034
![]() ![]() |
OriGene
TA503062 |
100 μL |
Each for $516.29
|
|
|||||
Description
ALDOB Monoclonal antibody specifically detects ALDOB in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
ALDOB | |
0.84 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
IgG1 | |
-20° C, Avoid Freeze/Thaw Cycles |
OTI 5D1 | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title